Skip to Content
Merck
All Photos(1)

Key Documents

AB1255

Sigma-Aldrich

Anti-Cytochrome P450 Enzyme CYP1A1 and CYP1A2 Antibody

serum, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
ELISA
WB
species reactivity:
rat
technique(s):
ELISA: suitable
western blot: suitable
citations:
3

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... AHRR(57491)

Specificity

Reacts with rat cytochrome P450 enzyme CYP1A1 and CYP1A2 on Western blots of hepatic microsomal fractions. No cross-reactivity with other cytochrome P450 enzymes.

Immunogen

Synthetic peptide

Application

Detect Cytochrome P450 Enzyme CYP1A1 & CYP1A2 using this Anti-Cytochrome P450 Enzyme CYP1A1 & CYP1A2 Antibody validated for use in ELISA & WB.
Research Category
Metabolism
Research Sub Category
Enzymes & Biochemistry
Western blot: useful for the simultaneous detection of rat CYP1A1 (57 kDa) and CYP1A2 (54 kDa), which can be separated electrophoretically.

Optimal working dilutions must be determined by end user.

Physical form

Rabbit antiserum. Liquid. Contains no preservative.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
POSITIVE CONTROL: rat hepatic microsomal fraction.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephanie E Martinez et al.
Drug metabolism and disposition: the biological fate of chemicals, 47(11), 1314-1324 (2019-08-21)
Dogs are commonly used in human and veterinary pharmaceutical development. Physiologically based pharmacokinetic modeling using recombinant cytochrome P450 (CYP) enzymes requires accurate estimates of CYP abundance, particularly in liver. However, such estimates are currently available for only seven CYPs, which
A role of Wnt/beta-catenin signals in hepatic fate specification of human umbilical cord blood-derived mesenchymal stem cells.
Yoshida, Y; Shimomura, T; Sakabe, T; Ishii, K; Gonda, K; Matsuoka, S; Watanabe, Y; Takubo et al.
American Journal of Physiology: Gastrointestinal and Liver Physiology null
Stephanie E Martinez et al.
Scientific reports, 10(1), 69-69 (2020-01-11)
Greyhounds recover more slowly from certain injectable anesthetics than other dog breeds. Previous studies implicate cytochrome P450 (CYP) 2B11 as an important clearance mechanism for these drugs and suggest Greyhounds are deficient in CYP2B11. However, no CYP2B11 gene mutations have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service