901782
Seal (O-ring 56x3.6 mm)
for use with withdrawl systems (products 1.01114 and 1.01123)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121516
Recommended Products
compatibility
for use with withdrawl systems (products 1.01114 and 1.01123)
shipped in
ambient
storage temp.
no temp limit
Related Categories
General description
If withdrawal system parts are lost or damaged, spare parts are available, e.g. special sealings for our withdrawal systems. This seal is the size of G2 and fits to the threaded adapters 9.67100.9002, 9.67100.9003 as well as to the withdrawal systems 1.01114.0001 and 1.01123.0001.
for withdrawal systems Cat.Nos. 1.01114.0001 and 1.01123.0001
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service