Skip to Content
Merck
All Photos(1)

Key Documents

579053

Sigma-Aldrich

TAPI-1

≥97% (HPLC), liquid, MMP and TACE inhibitor, Calbiochem®

Synonym(s):

InSolution TAPI-1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H37N5O5
Molecular Weight:
499.60
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

TAPI-1, InSolution, ≥97%, inhibits MMPs and TACE

Quality Level

Assay

≥97% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

shipped in

wet ice

storage temp.

−20°C

General description

A structural analog of TAPI-0 (Cat. No. 579050) with similar in vitro efficacy for the inhibition of MMPs and TACE (TNF-α convertase; ADAM17). However, TAPI-1 is more stable in serum than TAPI-0. TAPI-1 Blocks the shedding of several cell surface proteins such as TNF-α, IL-6 receptor, and p60 and p80 TNF receptors. Inhibits constitutive (IC50 = 8.09 μM) and muscarinic receptor-stimulated (IC50 = 3.61 μM) sAPPa release in HEK-293 cells expressing muscarinic receptors. Also blocks the TACE-dependent constitutive release of co-transfected APP(695) (IC50 = 920 nM).

Biochem/physiol Actions

Cell permeable: no
Primary Target
MMPs and TACE
Product does not compete with ATP.
Reversible: no
Target IC50: 8.09 µM, 3.61 µM in blocking constitutive and muscarinic receptor-stimulated sAAPα release in HEK 293 cells expressing M3 muscarinic receptors

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 10 mM (500 µg/100 µl) solution of TAPI-1 (Cat. No. 579051) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah Bui et al.
Methods in molecular biology (Clifton, N.J.), 2557, 453-493 (2022-12-14)
In this chapter, we provide a detailed guide for the application of commonly used small molecules to study Golgi structure and function in vitro. Furthermore, we have curated a concise, validated list of endomembrane markers typically used in downstream assays

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service