Skip to Content
Merck
All Photos(1)

Key Documents

5.05227

Sigma-Aldrich

(-)-Cytisine

Synonym(s):

(-)-Cytisine, nAChR Agonist, (-)-Cytisine, Nicotinic Acetylcholine Receptor Agonist, (-)-Cytisine, Baptitoxine, Sophorine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14N2O
CAS Number:
Molecular Weight:
190.24
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥99% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

water: 100 mM

storage temp.

2-8°C

InChI

1S/C11H14N2O/c14-11-3-1-2-10-9-4-8(5-12-6-9)7-13(10)11/h1-3,8-9,12H,4-7H2/t8-,9-/m0/s1

InChI key

ANJTVLIZGCUXLD-IUCAKERBSA-N

General description

A potent agonist selective for neuronal nAChRs (Ki = 2, 5890, 480, 329, and 492 nM at α4β2, α7, α3β4, α6/α4β4, α1β1γδ, respectively). Clinically used as a smoking cessation medicine and often used in addiction studies.

Biochem/physiol Actions

Primary Target
nAChR
Reversible: yes
Target Ki: 2, 5890, 480, 329, and 492 nM at &alpha

Warning

Toxicity: Toxic (F)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C. Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Chen, Y., et al., 2012. Br J Pharmacol.165, 1006.
West, R., 2011, N. Engl. J. Med.365, 1193.

Happe, K., et al., 1994. Neurosci.62, 929.

Reavill, C. et al., 1990. Neuropharmacol.29, 619.
Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service