Skip to Content
Merck
All Photos(1)

Key Documents

481975

Sigma-Aldrich

(--)-Nicotine, Ditartrate

Potent agonist of the nicotinic acetylcholine receptor.

Synonym(s):

(--)-Nicotine, Ditartrate, (S)-3-(1-Methyl-2-pyrrolindinyl)pyridine, Di- d-tartrate, (S)-3-(1-Methyl-2-pyrrolindinyl)pyridine, Di-d-tartrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2 · 2C4H6O6
CAS Number:
Molecular Weight:
462.41
MDL number:
UNSPSC Code:
12352210
NACRES:
NA.77

description

Merck USA index - 14, 6524

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to off-white

solubility

ethanol: 10 mg/mL
water: 10 mg/mL

shipped in

ambient

storage temp.

10-30°C

SMILES string

N1(C(CCC1)c2cnccc2)C.O[C@H]([C@@H](O)C(=O)O)C(=O)O.O[C@H]([C@@H](O)C(=O)O)C(=O)O

InChI

1S/C10H14N2.2C4H6O6/c1-12-7-3-5-10(12)9-4-2-6-11-8-9;2*5-1(3(7)8)2(6)4(9)10/h2,4,6,8,10H,3,5,7H2,1H3;2*1-2,5-6H,(H,7,8)(H,9,10)/t;2*1-,2-/m.11/s1

InChI key

RFEJUZJILGIRHQ-WBPXWQEISA-N

General description

Potent agonist of the nicotinic acetylcholine receptor. Stimulates autonomic ganglia and skeletal muscle neuromuscular junctions.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Nicotinic acetylcholine receptor
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Highly Toxic (H)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C) for long-term storage or refrigerate (4°C) for short-term storage. Stock solutions are stable for up to 2 weeks at 4°C or for up to 6 months at -20°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Rand, M.J. 1989. Prog. Brain Res. 79, 3.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

289.8 °F

Flash Point(C)

143.2 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yan Wang et al.
Oncology reports, 40(4), 2097-2104 (2018-07-18)
Store‑operated calcium entry (SOCE) is critical for regulating the proliferation and metastasis of various cancer types. The present study aimed to investigate the role of SOCE on nicotine‑promoted proliferation of non‑small cell lung cancer (NSCLC) A549 cells. Cell proliferation was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service