Skip to Content
Merck
All Photos(1)

Key Documents

219421

Sigma-Aldrich

Cathepsin L Inhibitor I

The Cathepsin L Inhibitor I, also referenced under CAS 108005-94-3, controls the biological activity of Cathepsin L. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

Cathepsin L Inhibitor I, Z-FF-FMK

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H27FN2O4
CAS Number:
Molecular Weight:
462.51
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white

solubility

DMSO: 20 mM

shipped in

ambient

storage temp.

−20°C

General description

A potent, cell-permeable and irreversible inhibitor of cathepsin L.
A potent, cell-permeable, and irreversible inhibitor of Cathepsin L (Cat. No. 219402).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
cathepsin L
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Sequence

Z-Phe-Phe-CH₂F

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Analysis Note

single spot by TLC

Other Notes

Ravanko, K., et al. 2004. Cancer Res.64, 8831.
Esser, R.E., et al. 1994. Arthritis Rheum. 37, 236.
Shaw, E., et al. 1993. FEBS Lett.334, 340.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael R Blatchley et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 8(14), e2100190-e2100190 (2021-06-22)
With the increased realization of the effect of oxygen (O2 ) deprivation (hypoxia) on cellular processes, recent efforts have focused on the development of engineered systems to control O2 concentrations and establish biomimetic O2 gradients to study and manipulate cellular
Hui Yang et al.
EMBO reports, 24(10), e57032-e57032 (2023-08-31)
Bromodomain-containing protein 4 (BRD4) is overexpressed and functionally implicated in various myeloid malignancies. However, the role of BRD4 in normal hematopoiesis remains largely unknown. Here, utilizing an inducible Brd4 knockout mouse model, we find that deletion of Brd4 (Brd4Δ/Δ )
Xiaoping Huang et al.
Virology journal, 20(1), 2-2 (2023-01-08)
Recent seminal studies have revealed that endosomal reactive oxygen species (ROS) promote rather than inhibit viral infection. Some ROS generators, including shikonin and H2O2, have the potential to enhance recombinant adeno-associated virus (rAAV) transduction. However, the impact of ROS on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service