Skip to Content
Merck
All Photos(1)

Documents

1.93009

Supelco

Boric acid

for analysis EMPARTA® ACS

Synonym(s):

Boric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3BO3
CAS Number:
Molecular Weight:
61.83
MDL number:
UNSPSC Code:
12352106

grade

ACS reagent

vapor pressure

2.6 mmHg ( 20 °C)

mp

160 °C (dec.) (lit.)

storage temp.

room temp

SMILES string

OB(O)O

InChI

1S/BH3O3/c2-1(3)4/h2-4H

InChI key

KGBXLFKZBHKPEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service