Skip to Content
Merck
All Photos(2)

Key Documents

1.10015

Supelco

Aluminum Test Strips, colorimetric

greener alternative

10-250 mg/L (Al), MQuant®

Synonym(s):

Aluminium chemical test strips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

Product Name

Aluminium Test, colorimetric, 10-250 mg/L (Al), MQuant®

description

semi-quantitative

Quality Level

product line

MQuant®

specific analyte(s)

aluminium

greener alternative product characteristics

Waste Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

measuring range

10-250 mg/L (Al)

detection method

colorimetric

greener alternative category

storage temp.

15-25°C

General description

Aluminium compounds are used as precipitating agents (flocculants) for water treatment plants, swimming baths and in paper manufacturing. MQuant Aluminium test strips can usually be applied directly here to test the effectiveness of the filter in process control. Using these test strips, the aluminium concentration is measured semiquantitatively by visual comparison of the reaction zone on the test strip with the fields of the color scale. All necessary reagents for the test are already included for a fast and easy analysis. The test strips are perfect for pre-testing a sample in an economic and quick way to estimate the aluminium concentration and a possible dilution factor prior to using a quantitative method such as a Reflectoquant and Spectroquant test.

Design for sustainability benefits:

We continue to demonstrate our commitment to our planet and a sustainable future by exploring ways to improve existing products in our portfolio. We have recently reviewed the current line of MQuant® test strips to identify key opportunities to minimize impact without compromising on the product performance.

This product has benefited from:

  • Test strip with reduction
  • Recycled aluminum tube
  • Digital instruction leaflet
Learn more about the initiative: Small Changes to Test Strips for Big Impact.
Method: colorimetric with test strips and reagents 10 - 25 - 50 - 100 - 250 mg/l Al MQuant®

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service