600430AVT
Avanti
250 mL Wide Mouth Packer
Avanti Research™ - A Croda Brand
Synonym(s):
250 mL amber wide mouth bottle with PTFE lined screw cap closure
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
24122000
NACRES:
NA.25
Recommended Products
manufacturer/tradename
Avanti Research™ - A Croda Brand
storage temp.
room temp
Related Categories
Packaging
pkg of 1 x 250 mL bottle (600430-1EA)
Legal Information
Avanti Research is a trademark of Avanti Polar Lipids, LLC
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service