Z678066
Finnpipette® F2 multichannel pipette
volume 30-300 μL, 12 channel
Synonym(s):
Multichannel Pipette
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
Recommended Products
feature
12-channel
color-coded orange
electronic: no
packaging
pack of 1 ea
manufacturer/tradename
Finn 4662070
volume
30-300 μL
Looking for similar products? Visit Product Comparison Guide
General description
Finnpipette F2 has a light pipetting action, is easy to use and is fully autoclavable. Accuracy and precision are the most vital aspect of any pipette so the Finnpipette F2 was developed with Advanced Volume Gearing (AVG). This is a self-supporting modular volume adjustment mechanism separated from the pipette body. This improves accuracy and precision by preventing possible effects of hand warmth on the accuracy of measurements.
- Advanced Volume Gearing mechanism (AVG) ensures increased accuracy and precision
- Big display with white numbers on black background for increased visibility
- Double action pipetting button with very light and smooth pipetting action
- Fully autoclavable
- Super blow-out piston with volumes of 50 μl and below
Legal Information
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service