Key Documents
Z630012
Sigma-Aldrich
IKA® FK 1 flexible coupling for A11 mill
FK 1 flexible coupling, 1/cs
About This Item
UNSPSC Code:
41101708
NACRES:
NB.72
Pricing and availability is not currently available. Contact Technical Service
feature
CE compliant
CSA approved
packaging
case of 1
pack of 1
manufacturer/tradename
IKA 23360.00
diam.
8 mm , clamping plug
size
6-10 mm , clamping range
General description
The flexible coupling is required for stirring jobs in glass apparatus. A torque of max. 10 Ncm can be tranfered to KPG-glass stirrers. The FK 1 balances inaccuracies of the set-up.
  • Required for stirring tasks using glass stirring rods
  • The flexible coupling compensates for any structural variances
Legal Information
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)