Z556203
Aldrich® Soxhlet extraction apparatus
extractor capacity 200 mL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
extractor capacity
200 mL
flask capacity
300 mL
thimble I.D. × H
43 mm × 123 mm
Looking for similar products? Visit Product Comparison Guide
General description
Soxhlet extraction apparatus is primarily used for the extraction of soil and sediment samples with mid-polar to nonpolar solvents.
Apparatus includes extractor body, condenser, and flask.
Application
Soxhlet extraction apparatus has been used for the extraction of Calotropis procera leaves with a solvent for determining the anti-inflammatory property of the shrub, which is used in folk medicine to treat several ailments.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
replacement part
Product No.
Description
Pricing
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Nanomaterials (Basel, Switzerland), 8(8) (2018-08-08)
This research deals with the effect of the temperature on the physical, thermal, electrochemical, and adsorption properties of the carbon micro-spheres using hydrothermal carbonization (HTC). Until recently, limited research has been conducted regarding the effects of delignification during the HTC
Phytochemical Screening and Anti-Inflammatory Properties of the Hydroethanolic Leaf Extract of Calotropis procera (Ait). R. Br. (Apocynaceae).
Journal of Pharmaceutical Research International, 23(1), 1-11 (2018)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service