Key Documents
Z549673
Sigma-Aldrich
High-vacuum valve adapter
male joint: ST/NS 14/20
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
Joint
male joint: ST/NS14/20
General description
For high-vacuum applications. Features a high-vacuum PTFE valve. For use with 1/4in. I.D. tubing.