Z547735
Aldrich® Buchner funnel with joint
capacity 30 mL, disc diam. 30 mm, porosity 10-20 μm, joint: ST/NS 14/20
Synonym(s):
buchner funnels, filter funnel, filter funnels
All Photos(1)
About This Item
Recommended Products
Joint
joint: ST/NS14/20
capacity
30 mL
disc diam.
30 mm
pore size
10-20 μm porosity
Related Categories
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)