Z408506
Clavies® biohazard autoclave gloves
gauntlet L 11 in. (28 cm), Overall L 18 1/2 in. (47 cm)
Synonym(s):
autoclave glove, heat shielding glove, oven glove
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
46181504
NACRES:
NB.35
Recommended Products
material
cotton
size
one size fits all
sterility
non-sterile
feature
CE compliant
latex free
powder-free
reusable
packaging
pkg of 1 pair
manufacturer/tradename
Bel-Art H13201-0001
Overall L
18 1/2 in. (47 cm)
gauntlet L
11 in. (28 cm)
color
orange
Looking for similar products? Visit Product Comparison Guide
General description
Soft, pliant, all-cotton terry cloth gloves are excellent for handling hot objects out of an autoclave or oven. Gauntlets available in two lengths and provide wrist and forearm protection.
Features and Benefits
- Heat-resistant up to 232°C (450°F).
- Can replace hazardous asbestos gloves.
- Provides wrist and forearm protection.
- Bright orange color offers high visibility.
- Machine wash, air dry
Legal Information
Clavies is a registered trademark of SP Scienceware, a division of SP
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service