Z202738
Jacketed beaker
Synonym(s):
Double-walled beaker, Insulated beaker
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41121803
NACRES:
NB.43
Recommended Products
material
borosilicate glass
feature
graduations: no
volume
50 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Outer OD - 45mm x H 120mm
Inner ID - ~27mm x D 110mm (inner OD - 30mm)
Inner ID - ~27mm x D 110mm (inner OD - 30mm)
One upper and one lower hose connection on opposite sides. Without pouring spout. A 3/8 in. I.D. hose is specified. A vacuum is not used with this product. Hot or cold liquids are circulated through the jacket to control temperature within the beaker.
Other Notes
Browse all glass beakers
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service