Key Documents
SYNF414450
Synthware two neck round bottom flask with angled side neck
50 mL, center joint: ST/NS 24/40, side joint: ST/NS 24/40
Synonym(s):
Synthware F414450, 2-neck round flask
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
material
borosilicate glass flask
round bottom
two-neck (angled side neck)
Joint
center joint: ST/NS24/40
side joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F414450
capacity
50 mL
50 mL
General description
Round bottom with joints. Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Legal Information
Synthware is a trademark of Kemtech America Inc.