GF82136190
Glass Sealing Alloy, Fe54Ni29Co17
foil, 4mm disks, 0.30mm thickness, annealed
Synonym(s):
FE040305
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
form
annealed
foil (disks)
manufacturer/tradename
Goodfellow 821-361-90
size × thickness
4 mm × 0.30 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service