GF77949590
Fecralloy® - iron/chromium
foil, thickness 0.05 mm, Fe72.8%/Cr22%/Al 5%/Y 0.1%/Zr 0.1%, size 250 × 250 mm, temper as rolled
Synonym(s):
FE080251
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
11101712
NACRES:
NA.23
Recommended Products
form
foil
manufacturer/tradename
Goodfellow 779-495-90
size
250 × 250 mm
thickness
0.05 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com
Legal Information
Product of Goodfellow
Fecralloy is a registered trademark of SANDVIK INTELLECTUAL PROPERTY AB
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service