CLS842450
Pyrex® round-bottom centrifuge tube
capacity 50 mL, O.D. × L 29 mm × 133 mm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121703
NACRES:
NB.43
Recommended Products
material
borosilicate glass
cap (#13 Pennyhead Stopper)
conical bottom tube
sterility
non-sterile
packaging
case of 12
pack of 12
manufacturer/tradename
Corning 8424-50
availability
not available in EU
O.D. × L
29 mm × 133 mm
capacity
50 mL
Related Categories
General description
With pennyhead stopper.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service