Key Documents
CLS70340500
Pyrex® VISTA filter flask, heavy-walled
capacity 500 mL
Synonym(s):
conical flasks, filtering flask, filtering flasks, flasks erlenmeyer, Corning® 70340
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
flat bottom flask
narrow-mouth
sterility
non-sterile
feature
closure type Rubber stoppers
packaging
case of 12
pack of 6
manufacturer/tradename
Corning 70340-500
capacity
500 mL
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
VISTA is a trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)