Skip to Content
Merck
All Photos(1)

Key Documents

C30000

Sigma-Aldrich

4-Chlorobutyronitrile

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl(CH2)3CN
CAS Number:
Molecular Weight:
103.55
Beilstein:
1633582
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.442 (lit.)

bp

195-197 °C (lit.)

density

1.095 g/mL at 20 °C

SMILES string

ClCCCC#N

InChI

1S/C4H6ClN/c5-3-1-2-4-6/h1-3H2

InChI key

ZFCFBWSVQWGOJJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

185.0 °F

Flash Point(C)

85 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service