BR4600210
BRAND® Dispensette® S Fixed-volume bottle-top dispenser
volume 1 mL, without recirculation valve
All Photos(1)
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
feature
DE-M marking
without recirculation valve
manufacturer/tradename
BRAND 4600210
volume
1 mL
fitting
(GL 28/S28)
GL 32
GL 38
S 40
General description
Dispensette S is suitable for an amazingly broad range of applications, particularly for many acids, saline solutions and organic solvents.
Features and Benefits
- Extremely easy filling and dispensing
- Simple priming by optimized flow channels
- Rapid and safe volume adjustment with internal scalloped track
Other Notes
Fixed Volume- For standard applications. Simple calibration adjustment with a supplied tool.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Related Content
Chemical compatibility chart for all Dispensette® S and Dispensette® S Organic models.