Merck
All Photos(5)
Key Documents
BAF371190083
SP Bel-Art® Spinbar® magnetic stirring fleas
white, L × diam. 0.315 in. (8 mm) × 0.118 in. (3 mm)
Synonym(s):
Spinbar® magnetic stirring fleas
About This Item
UNSPSC Code:
41122800
NACRES:
NB.51
material
PTFE-coated
white
feature
ALNICO V coated magnet
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F37119-0083
L × diam.
0.315 in. (8 mm) × 0.118 in. (3 mm)
General description
  • Spinbar® magnetic stirring fleas are designed for use in tubes, vials, and gradient markers.
  • Ideal for mixing of small volume samples in environmental testing and life science applications.
  • Alnico magnet encapsulated in durable and inert FDA grade PTFE (polytetrafluoroethylene) coating.
  • Broad chemical resistance for use in a wide variety of applications.
  • Autoclavable.
Designed for small volume mixing in vessels such as vials, tubes and gradient makers.
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Spinbar is a registered trademark of SP Scienceware, a division of SP
Teflon is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)