Skip to Content
Merck
All Photos(1)

Key Documents

B18200

Sigma-Aldrich

Benzyl carbamate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2COOCH2C6H5
CAS Number:
Molecular Weight:
151.16
Beilstein:
1865635
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

flakes

mp

86-89 °C (lit.)

SMILES string

NC(=O)OCc1ccccc1

InChI

1S/C8H9NO2/c9-8(10)11-6-7-4-2-1-3-5-7/h1-5H,6H2,(H2,9,10)

InChI key

PUJDIJCNWFYVJX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Shin Ono et al.
Biopolymers, 106(4), 521-530 (2015-12-01)
Diphenyl (α-aminoalkyl)phosphonates act as mechanism-based inhibitors against serine proteases by forming a covalent bond with the hydroxy group of the active center Ser residue. Because the covalent bond was found to be broken and replaced by 2-pyridinaldoxime methiodide (2PAM), we
Sevnur Serim et al.
Organic & biomolecular chemistry, 13(8), 2293-2299 (2015-01-03)
Activity-based probes (ABPs) are powerful tools for the analysis of active enzyme species in whole proteomes, cells or animals. Quenched fluorescent ABPs (qABPs) can be applied for real time imaging, allowing the visualization of dynamic enzyme activation by fluorescent microscopy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service