Skip to Content
Merck
All Photos(1)

Key Documents

927457

Sigma-Aldrich

SDASO-S crosslinker

≥95%

Synonym(s):

2,5-Dioxopyrrolidin-1-yl 3-((2-(3-methyl-3H-diazirin-3-yl)ethyl)sulfinyl)propanoate, Succinimidyl diazirine sulfoxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15N3O5S
Molecular Weight:
301.32
MDL number:
UNSPSC Code:
12352106

Quality Level

Assay

≥95%

form

powder

mp

87 °C

storage temp.

−20°C

SMILES string

O=C(CCS(CCC1(N=N1)C)=O)ON2C(CCC2=O)=O

InChI

1S/C11H15N3O5S/c1-11(12-13-11)5-7-20(18)6-4-10(17)19-14-8(15)2-3-9(14)16/h2-7H2,1H3

InChI key

MSHAOLYSQMKWAS-UHFFFAOYSA-N

Application

SDASO-S (succinimidyl diazirine sulfoxide) is mass-spectrometry cleavable heterobifunctional cross-linker, which can be used in the analysis of protein structure, or protein-protein interactions. SDASO-S is photoactivated, allowing for the first time photocross-linking of proteins, and rapid, accurate analysis by MS.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Other Notes

https://www.sciencedirect.com/science/article/pii/S1535947621000578">Enabling Photoactivated Cross-Linking Mass Spectrometric Analysis of Protein Complexes by Novel MS-Cleavable Cross-Linkers

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-react. C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Craig Gutierrez et al.
Molecular & cellular proteomics : MCP, 20, 100084-100084 (2021-04-30)
Cross-linking mass spectrometry (XL-MS) is a powerful tool for studying protein-protein interactions and elucidating architectures of protein complexes. While residue-specific XL-MS studies have been very successful, accessibility of interaction regions nontargetable by specific chemistries remain difficult. Photochemistry has shown great

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service