Skip to Content
Merck
All Photos(1)

Documents

858412

Sigma-Aldrich

(−)-2-Amino-6-mercaptopurine riboside hydrate

98%

Synonym(s):

Thioguanosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N5O4S · xH2O
CAS Number:
Molecular Weight:
299.31 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

optical activity

[α]22/D −69.9°, c = 1.3 in 0.1 M NaOH

mp

230-231 °C (dec.) (lit.)

SMILES string

[H]O[H].Nc1nc(S)c2ncn([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)c2n1

InChI

1S/C10H13N5O4S.H2O/c11-10-13-7-4(8(20)14-10)12-2-15(7)9-6(18)5(17)3(1-16)19-9;/h2-3,5-6,9,16-18H,1H2,(H3,11,13,14,20);1H2/t3-,5-,6-,9-;/m1./s1

InChI key

CEMWFPKZUAKCRS-GWTDSMLYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barbara Krystyna Stepien et al.
Proceedings of the National Academy of Sciences of the United States of America, 113(45), E7030-E7038 (2016-10-30)
Localized protein translation is critical in many biological contexts, particularly in highly polarized cells, such as neurons, to regulate gene expression in a spatiotemporal manner. The cytoplasmic polyadenylation element-binding (CPEB) family of RNA-binding proteins has emerged as a key regulator

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service