Skip to Content
Merck
All Photos(1)

Key Documents

769339

Sigma-Aldrich

Sodium hydrogen sulfate monohydrate

≥99% (T)

Synonym(s):

Sodium bisulfate monohydrate, Sodium hydrogen sulfate monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaHSO4 · H2O
CAS Number:
Molecular Weight:
138.08
EC Number:
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23
Assay:
≥99% (T)
form:
crystalline

Quality Level

Assay

≥99% (T)

form

crystalline

anion traces

chloride (Cl-): ≤0.003%

cation traces

Ca: ≤0.01%
Fe: ≤0.003%
heavy metals (as Pb): ≤0.004%

SMILES string

[Na+].[H]O[H].OS([O-])(=O)=O

InChI

1S/Na.H2O4S.H2O/c;1-5(2,3)4;/h;(H2,1,2,3,4);1H2/q+1;;/p-1

InChI key

JXHZRQHZVYDRGX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

The monohydrate of sodium hydrogen sulfate finds application as catalyst in organic reactions such as esterification of carboxylic acids and alcohols.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of thiol-functionalized spent grain as a novel adsorbent for divalent metal ions.
Chai, L., et al.
Bioresource Technology, 101(15), 6269-6272 (2010)
Shaterian, HR., et al.
Chin. J. Chem., 26, 338-342 (2008)
Catalytic esterifications of carboxylic acids and alcohols by sodium bisulfate monohydrate.
Li YQ.
Synthetic Communications, 29(22), 3901-3903 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service