Skip to Content
Merck
All Photos(1)

Key Documents

727423

Sigma-Aldrich

2-[2-(2-Azidoethoxy)ethoxy]ethanol solution

~0.5 M in tert-butyl methyl ether

Synonym(s):

8-Azido-3,6-dioxaoctanol solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13N3O3
CAS Number:
Molecular Weight:
175.19
Beilstein:
4418543
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (GC)

Quality Level

form

liquid

reaction suitability

reaction type: click chemistry
reagent type: linker

concentration

~0.5 M in tert-butyl methyl ether

impurities

≤1.5% water

functional group

azide
hydroxyl

storage temp.

2-8°C

SMILES string

OCCOCCOCCN=[N+]=[N-]

InChI

1S/C6H13N3O3/c7-9-8-1-3-11-5-6-12-4-2-10/h10H,1-6H2

InChI key

PMNIHDBMMDOUPD-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-27.4 °F - Not applicable

Flash Point(C)

-33 °C - Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of New Phospholipids Linked to Steroid-Hormone Derivatives Designed for Two-Dimensional Crystallization of Proteins
Lebeau, L., et al.
Helvetica Chimica Acta, 74, 1697-1706 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service