Skip to Content
Merck
All Photos(1)

Key Documents

717347

Sigma-Aldrich

1,4-Bis[(phenyl-3-propanesulfonate) phosphine] butane disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H30P2Na2S2O6
Molecular Weight:
562.53
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

form

solid

SMILES string

[Na+].[Na+].[O-]S(=O)(=O)CCCP(CCCCP(CCCS([O-])(=O)=O)c1ccccc1)c2ccccc2

InChI

1S/C22H32O6P2S2.2Na/c23-31(24,25)19-9-17-29(21-11-3-1-4-12-21)15-7-8-16-30(18-10-20-32(26,27)28)22-13-5-2-6-14-22;;/h1-6,11-14H,7-10,15-20H2,(H,23,24,25)(H,26,27,28);;/q;2*+1/p-2

InChI key

VCUCCQWAQVBMEN-UHFFFAOYSA-L

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jan-Bernd Hövener et al.
Angewandte Chemie (International ed. in English), 57(35), 11140-11162 (2018-02-28)
Magnetic resonance (MR) is one of the most versatile and useful physical effects used for human imaging, chemical analysis, and the elucidation of molecular structures. However, its full potential is rarely used, because only a small fraction of the nuclear

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service