Skip to Content
Merck
All Photos(2)

Documents

716251

Sigma-Aldrich

2,2,6,6-Tetramethyl-1,2,3,6-tetrahydro-4-pyridineboronic acid pinacol ester

97%

Synonym(s):

(2,2,6,6-Tetramethyl-3,6-dihydro-2H-pyridin-4-yl)boronic acid pinacol ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H28BNO2
CAS Number:
Molecular Weight:
265.20
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

95-100 °C

storage temp.

2-8°C

SMILES string

CC1(C)CC(=CC(C)(C)N1)B2OC(C)(C)C(C)(C)O2

InChI

1S/C15H28BNO2/c1-12(2)9-11(10-13(3,4)17-12)16-18-14(5,6)15(7,8)19-16/h9,17H,10H2,1-8H3

InChI key

YYVIRFZZNQUTKP-UHFFFAOYSA-N

General description

2,2,6,6-Tetramethyl-1,2,3,6-tetrahydro-4-pyridineboronic acid pinacol ester is a boronic ester that is commonly used as a reagent in Suzuki-Miyaura coupling reactions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service