Skip to Content
Merck
All Photos(1)

Key Documents

69830

Sigma-Aldrich

DL-Lactonitrile

≥97.0% (T)

Synonym(s):

2-Hydroxypropionitrile, Acetaldehyde cyanohydrin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CN
CAS Number:
Molecular Weight:
71.08
Beilstein:
605366
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (T)

form

liquid

contains

~1% phosphoric acid as stabilizer

impurities

~1% water

refractive index

n20/D 1.404

bp

90 °C/17 mmHg (lit.)

density

0.991 g/mL at 20 °C (lit.)

functional group

hydroxyl

SMILES string

CC(O)C#N

InChI

1S/C3H5NO/c1-3(5)2-4/h3,5H,1H3

InChI key

WOFDVDFSGLBFAC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 1 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Carc. 2 - Met. Corr. 1 - Repr. 1B - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R H Steele et al.
Archives of biochemistry and biophysics, 267(1), 125-142 (1988-11-15)
Cyanide added to mitochondria in the presence of copper and acetaldehyde evokes a chemiluminescence which follows series pseudo-first-order kinetics: (formula; see text) An evaluation of the effects of protein (mitochondria), copper, cyanide, acetaldehyde, and oxygen on the kinetic parameters shows
F Effenberger et al.
Current opinion in biotechnology, 11(6), 532-539 (2000-12-05)
(R)- as well as (S)-cyanohydrins are now easily available as a result of the excellent accessibility, the relatively high stability and the easy handling of hydroxynitrile lyases (HNLs). The optimization of reaction conditions (solvent, temperature, and using site-directed mutagenesis, etc.)
A D Keefe et al.
Origins of life and evolution of the biosphere : the journal of the International Society for the Study of the Origin of Life, 26(1), 15-25 (1996-02-01)
In view of the importance of a prebiotic source of high energy phosphates, we have investigated a number of potentially prebiotic processes to produce condensed phosphates from orthophosphate and cyclic trimetaphosphate from tripolyphosphate. The reagents investigated include polymerizing nitriles, acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service