Skip to Content
Merck
All Photos(1)

Key Documents

671592

Sigma-Aldrich

O-(2-Aminoethyl)-O′-(2-carboxyethyl)polyethylene glycol 5,000 hydrochloride

Mp 5,000

Synonym(s):

Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2O(CH2CH2O)nCH2CH2COOH . HCl
CAS Number:
UNSPSC Code:
51171641
NACRES:
NA.22

mol wt

Mp 5,000

reaction suitability

reagent type: cross-linking reagent

Ω-end

carboxylic acid

α-end

amine

Application

Used for:
  • Aggregation to vesicle polymersomes and immobilization of functionalized peroxidase of permethylated ß-cyclodextrin-appended polystyrene giant amphiphile
  • Platform for oligonucleotide delivery utilizing PEG prodrug approach
  • Folate-receptor-targeted delivery of doxorubicin nano-aggregates stabilized by doxorubicin-PEG-folate conjugate

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Progress in biotechnology fields such as tissue engineering and drug delivery is accompanied by an increasing demand for diverse functional biomaterials. One class of biomaterials that has been the subject of intense research interest is hydrogels, because they closely mimic the natural environment of cells, both chemically and physically and therefore can be used as support to grow cells. This article specifically discusses poly(ethylene glycol) (PEG) hydrogels, which are good for biological applications because they do not generally elicit an immune response. PEGs offer a readily available, easy to modify polymer for widespread use in hydrogel fabrication, including 2D and 3D scaffold for tissue culture. The degradable linkages also enable a variety of applications for release of therapeutic agents.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service