655139
Isovaleraldehyde-1-13C
99 atom % 13C, 97% (CP)
Synonym(s):
3-Methylbutanal-1-13C
About This Item
isotopic purity
99 atom % 13C
Assay
97% (CP)
bp
90 °C (lit.)
density
0.803 g/mL at 25 °C
mass shift
M+1
SMILES string
CC(C)C[13CH]=O
InChI
1S/C5H10O/c1-5(2)3-4-6/h4-5H,3H2,1-2H3/i4+1
InChI key
YGHRJJRRZDOVPD-AZXPZELESA-N
Related Categories
Packaging
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
23.0 °F - closed cup
Flash Point(C)
-5 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service