Skip to Content
Merck
All Photos(1)

Documents

487473

Sigma-Aldrich

1,4-Phenylenediamine-2,3,5,6-d4

98 atom % D

Synonym(s):

1,4-Benzene-d4-diamine, 1,4-Diaminobenzene-2,3,5,6-d4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6D4(NH2)2
Molecular Weight:
112.17
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

form

solid

bp

267 °C (lit.)

mp

143-145 °C (lit.)

mass shift

M+4

SMILES string

[2H]c1c([2H])c(N)c([2H])c([2H])c1N

InChI

1S/C6H8N2/c7-5-1-2-6(8)4-3-5/h1-4H,7-8H2/i1D,2D,3D,4D

InChI key

CBCKQZAAMUWICA-RHQRLBAQSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Sens. 1 - STOT SE 1 Oral

Target Organs

Kidney,Heart,Musculo-skeletal system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service