Skip to Content
Merck
All Photos(1)

Documents

441058

Sigma-Aldrich

(1R)-(−)-Menthyl acetate

98%

Synonym(s):

(-)-Menthol acetate, (-)-Menthyl acetate, (-)-p-Menthyl acetate, (1R,2S,5R)-5-Methyl-2-(1-methylethyl)cyclohexanol acetate, Acetic acid (R)-menthyl ester, L-Menthyl acetate, l-(-)-Menthyl acetate, l-Menthol acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O2
CAS Number:
Molecular Weight:
198.30
Beilstein:
2208505
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

optical activity

[α]20/D −81°, c = 8 in benzene

optical purity

ee: 98% (GLC)

refractive index

n20/D 1.447 (lit.)

bp

229-230 °C (lit.)

density

0.92 g/mL at 25 °C (lit.)

SMILES string

CC(C)[C@@H]1CC[C@@H](C)C[C@H]1OC(C)=O

InChI

1S/C12H22O2/c1-8(2)11-6-5-9(3)7-12(11)14-10(4)13/h8-9,11-12H,5-7H2,1-4H3/t9-,11+,12-/m1/s1

InChI key

XHXUANMFYXWVNG-ADEWGFFLSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • RIFM fragrance ingredient safety assessment, menthyl acetate (isomer unspecified): This safety assessment covers the toxicity data on menthyl acetate, indicating low toxicity and providing a foundation for safe usage in fragrances and related products (AM Api et al., 2017).

  • The crystalline sponge method: MOF terminal ligand effects: This article discusses the inclusion of (1R)-(−)-menthyl acetate into Metal-Organic Frameworks (MOFs) and analyzes its interactions and structural integration (TR Ramadhar et al., 2015).

  • Engineering a Bacillus subtilis esterase for selective hydrolysis of d, l-menthyl acetate in an organic solvent-free system: This research engineered a variant of Bacillus subtilis esterase that shows enhanced selectivity and efficiency in the hydrolysis of d, l-menthyl acetate, suitable for industrial applications (J Qiao et al., 2023).


Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service