401579
Lead(II) tetrafluoroborate solution
50 wt. % in H2O
Synonym(s):
Lead boron fluoride, Lead(II) fluoroborate
About This Item
Recommended Products
form
liquid
reaction suitability
core: lead
reagent type: catalyst
concentration
50 wt. % in H2O
density
1.615 g/mL at 25 °C
SMILES string
[PbH2++].F[B-](F)(F)F.F[B-](F)(F)F
InChI
1S/2BF4.Pb/c2*2-1(3,4)5;/q2*-1;+2
InChI key
UQGXAENNKXXYNF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Analyzed in terms of electochemical characterization of the effects of impurities and organic additives in lead electrowinning from fluoroborate electrolyte
Used in the development and optimization of a bipolar lead-boron tetrafluoride accumulator
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - STOT RE 2
Target Organs
Central nervous system,Reproductive organs
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service