Skip to Content
Merck
All Photos(1)

Key Documents

400610

Sigma-Aldrich

Samarium(III) chloride

anhydrous, powder, 99.9% trace rare earth metals basis

Synonym(s):

Samarium chloride, Samarium trichloride, Samarium(+3) chloride, Trichlorosamarium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SmCl3
CAS Number:
Molecular Weight:
256.72
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

grade

anhydrous

Assay

99.9% trace rare earth metals basis

form

powder

reaction suitability

reagent type: catalyst
core: samarium

impurities

≤1500.0 ppm Trace Rare Earth Analysis

mp

686 °C (lit.)

density

4.46 g/mL at 25 °C (lit.)

SMILES string

Cl[Sm](Cl)Cl

InChI

1S/3ClH.Sm/h3*1H;/q;;;+3/p-3

InChI key

BHXBZLPMVFUQBQ-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Markus Martincic et al.
Physical chemistry chemical physics : PCCP, 17(47), 31662-31669 (2015-11-12)
The endohedral functionalization of carbon nanotubes with both organic and inorganic materials allows the development of tailored functional hybrids whose properties benefit from the synergistic effects of the constituent compounds. Bulk filling of carbon nanotubes (CNTs) results in samples that

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service