Skip to Content
Merck
All Photos(3)

Key Documents

294144

Sigma-Aldrich

8-Bromo-1-octanol

95%

Synonym(s):

Octamethylene bromohydrin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH2)8OH
CAS Number:
Molecular Weight:
209.12
Beilstein:
1900837
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

reaction suitability

reagent type: cross-linking reagent

refractive index

n20/D 1.480 (lit.)

bp

79-80 °C/0.07 mmHg (lit.)

density

1.22 g/mL at 25 °C (lit.)

functional group

bromo
hydroxyl

storage temp.

2-8°C

SMILES string

BrCCCCCCCCO

InChI

1S/C8H17BrO/c9-7-5-3-1-2-4-6-8-10/h10H,1-8H2

InChI key

GMXIEASXPUEOTG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neha Kaushal et al.
Journal of controlled release : official journal of the Controlled Release Society, 289, 79-93 (2018-08-28)
Overexpression of RhoC protein in breast cancer patients has been linked to increased cancer cell invasion, migration, and metastases. Suppressing RhoC expression in aggressive breast cancer cells using silencing RNA (siRNA) molecules is a viable strategy to inhibit the metastatic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service