All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C8H6N2O2
CAS Number:
Molecular Weight:
162.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
mp
205-207 °C (lit.)
SMILES string
[O-][N+](=O)c1cccc2[nH]ccc12
InChI
1S/C8H6N2O2/c11-10(12)8-3-1-2-7-6(8)4-5-9-7/h1-5,9H
InChI key
LAVZKLJDKGRZJG-UHFFFAOYSA-N
Related Categories
Application
4-Nitroindole was used in the synthesis of 1,3,4,5-tetrahydropyrrolo-[4,3,2-de]quinoline.
Reactant for preparation of:
- Tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators
- Meridianin derivatives as protein kinase (PKC) inhibitors and vitro antiproliferative agents
- Anti-angiogenic agents
- Sodium-dependent glucose co-transporter 2 (SGLT2) inhibitors for the management of hyperglycemia in diabetes
- Biologically active indoles
- CGRP receptor antagonists
- Nucleosides
- CB2 cannabinoid receptor ligands
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Efficient Synthesis of 1, 3, 4, 5-Tetrahydropyrrolo-[4, 3, 2-de] Quinoline.
Synthetic Communications, 25(5), 681-690 (1995)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service