Skip to Content
Merck
All Photos(3)

Documents

155373

Sigma-Aldrich

Bis(2,2,2-trichloroethyl) phosphorochloridate

98%

Synonym(s):

Bis(2,2,2-trichloroethyl) chlorophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(Cl3CCH2O)2P(O)Cl
CAS Number:
Molecular Weight:
379.22
Beilstein:
1820707
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

mp

45-47 °C (lit.)

SMILES string

ClC(Cl)(Cl)COP(Cl)(=O)OCC(Cl)(Cl)Cl

InChI

1S/C4H4Cl7O3P/c5-3(6,7)1-13-15(11,12)14-2-4(8,9)10/h1-2H2

InChI key

ZHHCWQGVXYGWCW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Bis(2,2,2-trichloroethyl) phosphorochloridate was used in the preparation of epimeric 6-deoxyhexofuranosyl nucleoside 5′ -phosphate.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel analogs of nucleoside 3',5'-cyclic phosphates. I. 5'-Mono- and dimethyl analogs of adenosine 3',5'-cyclic phosphate.
R S Ranganathan et al.
The Journal of organic chemistry, 39(3), 290-302 (1974-02-08)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service