Merck
Long-Term Dexamethasone Exposure Down-Regulates Hepatic TFR1 and Reduces Liver Iron Concentration in Rats.
Nutrients (2017-06-21)
Huifang Li, Shuxia Jiang, Chun Yang, Shu Yang, Bin He, Wenqiang Ma, Ruqian Zhao
RESUMEN
Exposure to stress is known to cause hepatic iron dysregulation, but the relationship between prolonged stress and liver iron metabolism is not yet fully understood. Thirty 13-week-old female Sprague-Dawley rats were randomly divided into two groups, as follows: the control group (saline-injection) and the dexamethasone group (Dexamethasone (Dex)-injection 0.1 mg/kg/day). After the 21-day stress trial, the results showed that chronic Dex administration not only impaired serum corticosterone (
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-TF antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution