Merck
Investigating the Distribution of Stable Paramagnetic Species in an Apple Seed Using X-Band EPR and EPR Imaging.
Journal of oleo science (2017-02-14)
Kouichi Nakagawa, Boris Epel
RESUMEN
This study investigated the location and distribution of paramagnetic species in apple seeds using electron paramagnetic resonance (EPR) and X-band (9 GHz) EPR imaging (EPRI). EPR primarily detected two paramagnetic species per measured seed. These two different radical species were assigned as stable radicals and Mn
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
2,2,6,6-Tetramethyl-4-piperidinol, 98%