Merck
Diuretic-sensitive electroneutral Na
The Journal of physiology (2017-02-19)
Meneka Kanagaratnam, Christopher Pendleton, Danilo Almeida Souza, Joseph Pettit, James Howells, Mark D Baker
RESUMEN
Optic nerve axons get less excitable with warming. F-fibre latency does not shorten at temperatures above 30°C. Action potential amplitude falls when the Na Raising the temperature of optic nerve from room temperature to near physiological has effects on the threshold, refractoriness and superexcitability of the shortest latency (fast, F) nerve fibres, consistent with hyperpolarization. The temperature dependence of peak impulse latency was weakened at temperatures above 30°C suggesting a temperature-sensitive process that slows impulse propagation. The amplitude of the supramaximal compound action potential gets larger on warming, whereas in the presence of bumetanide and amiloride (blockers of electroneutral Na
MATERIALES
Referencia del producto
Marca
Descripción del producto
Ouabain, European Pharmacopoeia (EP) Reference Standard