Merck
Acoustic Emission from Organic Martensites.
Angewandte Chemie (International ed. in English) (2017-05-26)
Manas K Panda, Martin Etter, Robert E Dinnebier, Panče Naumov
RESUMEN
In salient effects, still crystals of solids that switch between phases acquire a momentum and are autonomously propelled because of rapid release of elastic energy accrued during a latent structural transition induced by heat, light, or mechanical stimulation. When mechanical reconfiguration is induced by change of temperature in thermosalient crystals, bursts of detectable acoustic waves are generated prior to self-actuation. These observations provide compelling evidence that the thermosalient transitions in organic and organic-containing crystals are molecular analogues of the martensitic transitions in some metals, and metal alloys such as steel and shape-memory alloys. Within a broader context, these results reveal that, akin to metallic bonding, the intermolecular interactions in molecular solids are capable of gradual accrual and sudden release of a substantial amount of strain during anisotropic thermal expansion, followed by a rapid transformation of the crystal packing in a diffusionless, non-displacive transition.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
1,2,4,5-Tetrabromobenzene, 97%