Merck
TRPM2 cation channels modulate T cell effector functions and contribute to autoimmune CNS inflammation.
PloS one (2012-10-19)
Nico Melzer, Gordon Hicking, Kerstin Göbel, Heinz Wiendl
RESUMEN
TRPM2, a highly Ca(2+)-permeable member of the transient receptor potential melastatin-related (TRPM) family of cation channels, is expressed in cells of the immune system. We demonstrate firstly that TRPM2 cation channels on T cells critically influence T cell proliferation and proinflammatory cytokine secretion following polyclonal T cell receptor stimulation. Consistently, trpm2-deficient mice exhibited an attenuated clincal phenotype of experimental autoimmune encephalomyelitis (EAE) with reduced inflammatory and demyelinating spinal cord lesions. Importantly, trmp2-deficient T cells were as susceptible as wildtype T cells to oxidative stress-induced cell death as it occurs in inflammatory CNS lesions. This supports the notion that the attenuated EAE phenotype is mainly due to reduced T cell effector functions but unaffected by potential modulation of T cell survival at the site of inflammation. Our findings suggest TRPM2 cation channels as a potential target for treating autoimmune CNS inflammation.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
N-(p-Amylcinnamoyl)anthranilic acid, ≥98% (HPLC)