Merck
GASDERMIN, suppressed frequently in gastric cancer, is a target of LMO1 in TGF-beta-dependent apoptotic signalling.
Oncogene (2007-05-02)
N Saeki, D H Kim, T Usui, K Aoyagi, T Tatsuta, K Aoki, K Yanagihara, M Tamura, H Mizushima, H Sakamoto, K Ogawa, M Ohki, T Shiroishi, T Yoshida, H Sasaki
RESUMEN
Defining apoptosis-regulatory cascades of the epithelium is important for understanding carcinogenesis, since cancer cells are considered to arise as a result of the collapse of the cascades. We previously reported that a novel gene GASDERMIN (GSDM) is expressed in the stomach but suppressed in gastric cancer cell lines. Furthermore, in this study, we demonstrated that GSDM is expressed in the mucus-secreting pit cells of the gastric epithelium and frequently silenced in primary gastric cancers. We found that GSDM has a highly apoptotic activity and its expression is regulated by a transcription factor LIM domain only 1 (LMO1) through a sequence to which Runt-related transcription factor 3 (RUNX3) binds, in a GSDM promoter region. We observed coexpression of GSDM with LMO1, RUNX3 and type II transforming growth factor-beta receptor (TGF-betaRII) in the pit cells, and found that TGF-beta upregulates the LMO1- and GSDM-expression in the gastric epithelial cell line and induces apoptosis, which was confirmed by the finding that the apoptosis induction is inhibited by suppression of each LMO1-, RUNX3- and GSDM expression, respectively. The present data suggest that TGF-beta, LMO1, possibly RUNX3, and GSDM form a regulatory pathway for directing the pit cells to apoptosis.