Merck
One-pot multienzyme synthesis of Lewis x and sialyl Lewis x antigens.
Current protocols in chemical biology (2012-09-01)
Hai Yu, Kam Lau, Yanhong Li, Go Sugiarto, Xi Chen
RESUMEN
L-Fucose has been found abundantly in human milk oligosaccharides, bacterial lipopolysaccharides, glycolipids, and many