Merck
  • Identification of a novel m.9588G > a missense mutation in the mitochondrial COIII gene in asthenozoospermic Tunisian infertile men.
Identification of a novel m.9588G > a missense mutation in the mitochondrial COIII gene in asthenozoospermic Tunisian infertile men.
Journal of assisted reproduction and genetics (2014-02-20)
Siwar Baklouti-Gargouri, Myriam Ghorbel, Afif Ben Mahmoud, Emna Mkaouar-Rebai, Meriam Cherif, Nozha Chakroun, Afifa Sellami, Faiza Fakhfakh, Leila Ammar-Keskes
RESUMEN
Infertility affects 10-15 % of the population, of which, approximately 40 % is due to male etiology consisting primarily of low sperm count (oligozoospermia) and/or abnormal sperm motility (asthenozoospermia). It has been demonstrated that mtDNA base substitutions can greatly influence semen quality. In the present study we performed a systematic sequence analysis of the mitochondrial cytochrome oxidase III (COIII) gene in 31 asthenozoospermic infertile men in comparaison to normozoospermic infertile men (n=33) and fertile men (n=150) from Tunisian population. A novel m.9588G>A mutation was found in the mtDNA sperm's in all asthenozoospermic patients and was absent in the normozoospermic and in fertile men. The m.9588G>A mutation substitutes a highly conserved Glutamate at position 128 to Lysine. In addition, PolyPhen-2 analysis predicted that this variant is "probably damaging".