Merck
  • Pyruvate dehydrogenase phosphatase deficiency: identification of the first mutation in two brothers and restoration of activity by protein complementation.
Pyruvate dehydrogenase phosphatase deficiency: identification of the first mutation in two brothers and restoration of activity by protein complementation.
The Journal of clinical endocrinology and metabolism (2005-04-28)
Mary C Maj, Neviana MacKay, Valeriy Levandovskiy, Jane Addis, E Regula Baumgartner, Matthias R Baumgartner, Brian H Robinson, Jessie M Cameron
RESUMEN
Pyruvate dehydrogenase phosphatase (PDP) deficiency has been previously reported as an enzymopathy, but the genetic basis for such a defect has never been established. The aim of this study was to identify the cause of the defect in two patients who presented with PDP deficiency. We studied two brothers of consanguineous parents who presented with neonatal hypotonia, elevated lactate, and less than 25% native pyruvate dehydrogenase complex (PDHc) activity in skin fibroblasts compared with controls. The activity of the complex could be restored to normal values by preincubation of the cells with dichloroacetate or by treating cell extracts with calcium. These two individuals were found to be homozygous for a 3-bp deletion in the coding sequence of the PDP isoform 1 (PDP1), which removes the amino acid residue leucine from position 213 of the protein. A recombinant version of this protein was synthesized and found to have a very reduced (<5%) ability to activate purified PDHc. Reduced steady-state levels of PDP1 in the patient's fibroblasts coupled with the low catalytic activity of the mutant PDP1 resulted in native PDHc activity being reduced, but this could be corrected by the addition of recombinant PDP1 (wild type). We have identified mutations in PDP1 in two brothers with PDP deficiency and have proven that the mutation is disease-causing. This is the first demonstration of human disease due to a mutation in PDP1.